gRNA Design Details

Target DNA Sequence
TCGACGGCCACTTGTATTTT
PAM Sequence
GGG
Target locus
Chr.2: 73828628 - 73828650 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 91.55/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Atf2
Entrez Gene ID
11909
Gene Name
activating transcription factor 2
Gene Aliases
Atf-2, CRE-BP, Creb2, D130078H02Rik, D18875, Tg(Gzma-Klra1)7Wum, mXBP
Chromosome Location
Chr.2: 73816509 - 73892639 on GRCm38
UniGene ID
Mm.209903
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001025093.2 NP_001020264.1 13
NM_001284369.1 NP_001271298.1 11
NM_001284370.1 NP_001271299.1 13
NM_001284371.1 NP_001271300.1 12
NM_001284372.1 NP_001271301.1 11
NM_001284373.1 NP_001271302.1 10
NM_001284374.1 NP_001271303.1 10
NM_001284376.1 NP_001271305.1 13
NM_009715.3 NP_033845.1 12