gRNA Design Details

Target DNA Sequence
GGCTCGTATACTGCCTCACT
PAM Sequence
CGG
Target locus
Chr.5: 145136949 - 145136971 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 86.37/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Pdap1
Entrez Gene ID
231887
Gene Name
PDGFA associated protein 1
Gene Aliases
HASPP28, PAP, PAP1
Chromosome Location
Chr.5: 145128770 - 145140089 on GRCm38
UniGene ID
Mm.188851
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001033313.3 NP_001028485.1 2