gRNA Design Details
- Target DNA Sequence
- GTACAGTTCTGCAGGAATGG
- PAM Sequence
- CGG
- Target locus
-
Chr.16: 56778644 - 56778666 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.21/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgrg7
- Entrez Gene ID
-
239853
- Gene Name
- adhesion G protein-coupled receptor G7
- Gene Aliases
-
9130020O16Rik, Gpr128
- Chromosome Location
-
Chr.16: 56724609 - 56796021 on GRCm38
- UniGene ID
- Mm.258979
- Species
-
Mouse