gRNA Design Details
- Target DNA Sequence
- TCGCGGTCATATCGGTATCC
- PAM Sequence
- CGG
- Target locus
-
Chr.18: 66084773 - 66084795 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
92.64/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ccbe1
- Entrez Gene ID
-
320924
- Gene Name
- collagen and calcium binding EGF domains 1
- Gene Aliases
-
4933426F18Rik, 9430093N24Rik, mKIAA1983
- Chromosome Location
-
Chr.18: 66056855 - 66291838 on GRCm38
- UniGene ID
- Mm.442049
- Species
-
Mouse