gRNA Design Details

Target DNA Sequence
GGCATTGATGCCCAGCTTCT
PAM Sequence
CGG
Target locus
Chr.4: 9844809 - 9844831 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 84.76/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Gdf6
Entrez Gene ID
242316
Gene Name
growth differentiation factor 6
Gene Aliases
BMP-13, BMP13, GDF16
Chromosome Location
Chr.4: 9844372 - 9862345 on GRCm38
UniGene ID
Mm.302555
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_013526.1 NP_038554.1 1