gRNA Design Details
- Target DNA Sequence
- GGTATGTTTACAATGGGGCA
- PAM Sequence
- AGG
- Target locus
-
Chr.18: 62594300 - 62594322 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spink7
- Entrez Gene ID
-
408198
- Gene Name
- serine peptidase inhibitor, Kazal type 7 (putative)
- Gene Aliases
-
EG408198, Ecg2
- Chromosome Location
-
Chr.18: 62592413 - 62596264 on GRCm38
- UniGene ID
- Mm.478742
- Species
-
Mouse