gRNA Design Details
- Target DNA Sequence
- AACTTCACTGGAATGCATCT
- PAM Sequence
- TGG
- Target locus
-
Chr.13: 52898693 - 52898715 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.74/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Auh
- Entrez Gene ID
-
11992
- Gene Name
- AU RNA binding protein/enoyl-coenzyme A hydratase
- Gene Aliases
-
C77140, W91705
- Chromosome Location
-
Chr.13: 52835102 - 52929706 on GRCm38
- UniGene ID
- Mm.252034
- Species
-
Mouse