gRNA Design Details
- Target DNA Sequence
- CTCCCCATTGGTGACTCTCG
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 30354911 - 30354933 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
83.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sh3glb2
- Entrez Gene ID
-
227700
- Gene Name
- SH3-domain GRB2-like endophilin B2
- Gene Aliases
-
SPAS-1
- Chromosome Location
-
Chr.2: 30344777 - 30359316 on GRCm38
- UniGene ID
- Mm.33343
- Species
-
Mouse