gRNA Design Details

Target DNA Sequence
CAACATGAAGAAGCTGGCGT
PAM Sequence
CGG
Target locus
Chr.9: 129028124 - 129028146 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 66.3/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
SH3GLB2
Entrez Gene ID
56904
Gene Name
SH3 domain containing GRB2 like endophilin B2
Gene Aliases
PP6569, PP9455, RRIG1
Chromosome Location
Chr.9: 129007792 - 129028353 on GRCh38
UniGene ID
Hs.460238
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001287045.1 NP_001273974.1 1
NM_001287046.1 NP_001273975.1 1
NM_020145.3 NP_064530.1 1
XM_005252098.2 XP_005252155.1 1
XM_005252100.2 XP_005252157.1 1
XM_005252101.3 XP_005252158.1 1
XM_006717188.2 XP_006717251.1 1
XM_006717189.2 XP_006717252.1 1
XM_006717190.1 XP_006717253.1 1
XM_006717191.1 XP_006717254.1 1