gRNA Design Details
- Target DNA Sequence
- TTGAGTCCACACTTTGCCCT
- PAM Sequence
- TGG
- Target locus
-
Chr.12: 15669721 - 15669743 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
67.65/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- EPS8
- Entrez Gene ID
-
2059
- Gene Name
- epidermal growth factor receptor pathway substrate 8
- Gene Aliases
-
DFNB102
- Chromosome Location
-
Chr.12: 15620141 - 15789576 on GRCh38
- UniGene ID
- Hs.591160
- Species
-
Human