gRNA Design Details
- Target DNA Sequence
- ACTACCTCCCGGCTGATGTT
- PAM Sequence
- GGG
- Target locus
-
Chr.10: 128891393 - 128891415 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
80.59/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Gdf11
- Entrez Gene ID
-
14561
- Gene Name
- growth differentiation factor 11
- Gene Aliases
-
BMP-11, Bmp11
- Chromosome Location
-
Chr.10: 128884546 - 128891718 on GRCm38
- UniGene ID
- Mm.299218
- Species
-
Mouse