gRNA Design Details

Target DNA Sequence
GCCCGTGCATCAATGTACGC
PAM Sequence
AGG
Target locus
Chr.11: 99041763 - 99041785 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 89.38/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Igfbp4
Entrez Gene ID
16010
Gene Name
insulin-like growth factor binding protein 4
Gene Aliases
AI875747, Deb2, IGFBP-4
Chromosome Location
Chr.11: 99041260 - 99052646 on GRCm38
UniGene ID
Mm.233799
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_010517.3 NP_034647.1 1