gRNA Design Details

Target DNA Sequence
ATTCGGAGGGCACCTCAGAC
PAM Sequence
AGG
Target locus
Chr.10: 87913645 - 87913667 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 86.98/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Igf1
Entrez Gene ID
16000
Gene Name
insulin-like growth factor 1
Gene Aliases
C730016P09Rik, Igf-1, Igf-I
Chromosome Location
Chr.10: 87858270 - 87937047 on GRCm38
UniGene ID
Mm.268521
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001111274.1 NP_001104744.1 3
NM_001111275.1 NP_001104745.1 3
NM_001111276.1 NP_001104746.1 3
NM_010512.4 NP_034642.2 3