gRNA Design Details
- Target DNA Sequence
- GTCATCATAGCTGCAGGTAC
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 24880067 - 24880089 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.38/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igfals
- Entrez Gene ID
-
16005
- Gene Name
- insulin-like growth factor binding protein, acid labile subunit
- Gene Aliases
-
ALS, Albs, mKIAA4111
- Chromosome Location
-
Chr.17: 24877653 - 24882011 on GRCm38
- UniGene ID
- Mm.3135
- Species
-
Mouse