gRNA Design Details

Target DNA Sequence
GGGTTTTGATGCAGTCACCG
PAM Sequence
GGG
Application
Gene Knockout
Score: 86.82/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Pak3
Entrez Gene ID
18481
Gene Name
p21 protein (Cdc42/Rac)-activated kinase 3
Gene Aliases
PAK-3, Pak65alpha, Pak65beta, Stk4
Chromosome Location
Chr.X: 143518368 - 143797796 on GRCm38
UniGene ID
Mm.40035
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001195046.1 NP_001181975.1 2
NM_001195047.1 NP_001181976.1 2
NM_001195048.1 NP_001181977.1 3
NM_001195049.1 NP_001181978.1 6
NM_008778.3 NP_032804.2 3