gRNA Design Details

Target DNA Sequence
GGACTCACCTGCATCTAGCA
PAM Sequence
GGG
Target locus
Chr.17: 56019145 - 56019167 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 84.89/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Sh3gl1
Entrez Gene ID
20405
Gene Name
SH3-domain GRB2-like 1
Gene Aliases
C77078, EEN, SH3P8, Sh3d2b
Chromosome Location
Chr.17: 56016750 - 56036638 on GRCm38
UniGene ID
Mm.1773
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001252471.1 NP_001239400.1 5
NM_013664.3 NP_038692.1 5