gRNA Design Details
- Target DNA Sequence
- GCGATTCAGGATTAGGAGGA
- PAM Sequence
- TGG
- Target locus
-
Chr.1: 51421921 - 51421943 on GRCh38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
65.77/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- EPS15
- Entrez Gene ID
-
2060
- Gene Name
- epidermal growth factor receptor pathway substrate 15
- Gene Aliases
-
AF-1P, AF1P, MLLT5
- Chromosome Location
-
Chr.1: 51354263 - 51519354 on GRCh38
- UniGene ID
- Hs.83722
- Species
-
Human