gRNA Design Details

Target DNA Sequence
GCGATTCAGGATTAGGAGGA
PAM Sequence
TGG
Target locus
Chr.1: 51421921 - 51421943 on GRCh38
Strand
Reverse
Application
Gene Knock-Out
Score: 65.77/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
EPS15
Entrez Gene ID
2060
Gene Name
epidermal growth factor receptor pathway substrate 15
Gene Aliases
AF-1P, AF1P, MLLT5
Chromosome Location
Chr.1: 51354263 - 51519354 on GRCh38
UniGene ID
Hs.83722
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001159969.1 NP_001153441.1 1