gRNA Design Details
- Target DNA Sequence
- TTTCAACGTGAAGAAGCTGG
- PAM Sequence
- CGG
- Target locus
-
Chr.3: 144720017 - 144720039 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
84.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sh3glb1
- Entrez Gene ID
-
54673
- Gene Name
- SH3-domain GRB2-like B1 (endophilin)
- Gene Aliases
-
AA409932, AI314629, AU015566, Bif-1, mKIAA0491
- Chromosome Location
-
Chr.3: 144683678 - 144720398 on GRCm38
- UniGene ID
- Mm.271775
- Species
-
Mouse