gRNA Design Details

Target DNA Sequence
GATTCTCTTCCAGGCAGCGC
PAM Sequence
TGG
Application
Gene Knock-Out
Score: 86.79/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Ikbkg
Entrez Gene ID
16151
Gene Name
inhibitor of kappaB kinase gamma
Gene Aliases
1110037D23Rik, AI848108, AI851264, AW124339, IKK[g], NEMO
Chromosome Location
Chr.X: 74393291 - 74456792 on GRCm38
UniGene ID
Mm.12967
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001136067.2 NP_001129539.1 3
NM_001161421.1 NP_001154893.1 3
NM_001161422.1 NP_001154894.1 2
NM_001161423.1 NP_001154895.1 2
NM_001161424.1 NP_001154896.1 2
NM_010547.2 NP_034677.2 2
NM_178590.4 NP_848705.1 2