gRNA Design Details

Target DNA Sequence
CTGTCCGTATAAGAGTGGTC
PAM Sequence
AGG
Target locus
Chr.15: 54259986 - 54260008 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 88.23/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Tnfrsf11b
Entrez Gene ID
18383
Gene Name
tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin)
Gene Aliases
OCIF, Opg, TR1
Chromosome Location
Chr.15: 54250619 - 54278484 on GRCm38
UniGene ID
Mm.15383
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_008764.3 NP_032790.3 2