gRNA Design Details
- Target DNA Sequence
- CTGTCCGTATAAGAGTGGTC
- PAM Sequence
- AGG
- Target locus
-
Chr.15: 54259986 - 54260008 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.23/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tnfrsf11b
- Entrez Gene ID
-
18383
- Gene Name
- tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin)
- Gene Aliases
-
OCIF, Opg, TR1
- Chromosome Location
-
Chr.15: 54250619 - 54278484 on GRCm38
- UniGene ID
- Mm.15383
- Species
-
Mouse