gRNA Design Details
- Target DNA Sequence
- TTTCGTTGAGGCAAACCCCG
- PAM Sequence
- CGG
- Target locus
-
Chr.2: 216694466 - 216694488 on GRCh38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
73.18/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- IGFBP5
- Entrez Gene ID
-
3488
- Gene Name
- insulin like growth factor binding protein 5
- Gene Aliases
-
IBP5
- Chromosome Location
-
Chr.2: 216672105 - 216695549 on GRCh38
- UniGene ID
- Hs.607212
- Species
-
Human