gRNA Design Details

Target DNA Sequence
TGCCTCAACGAAAAGAGCTA
PAM Sequence
CGG
Target locus
Chr.1: 72873823 - 72873845 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 85.7/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Igfbp5
Entrez Gene ID
16011
Gene Name
insulin-like growth factor binding protein 5
Gene Aliases
AI256729, AW208790, IGFBP-5, IGFBP-5P
Chromosome Location
Chr.1: 72858065 - 72874865 on GRCm38
UniGene ID
Mm.405761
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_010518.2 NP_034648.2 1