gRNA Design Details
- Target DNA Sequence
- TGCCTCAACGAAAAGAGCTA
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 72873823 - 72873845 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igfbp5
- Entrez Gene ID
-
16011
- Gene Name
- insulin-like growth factor binding protein 5
- Gene Aliases
-
AI256729, AW208790, IGFBP-5, IGFBP-5P
- Chromosome Location
-
Chr.1: 72858065 - 72874865 on GRCm38
- UniGene ID
- Mm.405761
- Species
-
Mouse