gRNA Design Details
- Target DNA Sequence
- TTGGACTAGGCCATGGTCCT
- PAM Sequence
- GGG
- Target locus
-
Chr.13: 22009465 - 22009487 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss16
- Entrez Gene ID
-
54373
- Gene Name
- protease, serine 16 (thymus)
- Gene Aliases
-
AI448615, TSSP
- Chromosome Location
-
Chr.13: 22002176 - 22009741 on GRCm38
- UniGene ID
- Mm.32043
- Species
-
Mouse