gRNA Design Details

Target DNA Sequence
GATCTGTCCCTTCCAAGTCC
PAM Sequence
TGG
Target locus
Chr.18: 36515102 - 36515124 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 85.9/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Hbegf
Entrez Gene ID
15200
Gene Name
heparin-binding EGF-like growth factor
Gene Aliases
AW047313, Dtr, Dts, Hegfl
Chromosome Location
Chr.18: 36504927 - 36515805 on GRCm38
UniGene ID
Mm.289681
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_010415.2 NP_034545.1 2