gRNA Design Details
- Target DNA Sequence
- AAGACGCCTACCAAATCCGA
- PAM Sequence
- GGG
- Target locus
-
Chr.4: 118486442 - 118486464 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tie1
- Entrez Gene ID
-
21846
- Gene Name
- tyrosine kinase with immunoglobulin-like and EGF-like domains 1
- Gene Aliases
-
D430008P04Rik, TIE, tie-1
- Chromosome Location
-
Chr.4: 118471191 - 118489849 on GRCm38
- UniGene ID
- Mm.4345
- Species
-
Mouse