gRNA Design Details

Target DNA Sequence
GTCCCATCAATGGTACCATC
PAM Sequence
TGG
Target locus
Chr.3: 192360451 - 192360473 on GRCh38
Strand
Forward
Application
Gene Knockout
Score: 71.78/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
FGF12
Entrez Gene ID
2257
Gene Name
fibroblast growth factor 12
Gene Aliases
FGF12B, FHF1
Chromosome Location
Chr.3: 192139393 - 192727599 on GRCh38
UniGene ID
Hs.390250
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_004113.5 NP_004104.3 3
NM_021032.4 NP_066360.1 2
XM_005247227.2 XP_005247284.1 2
XM_006713538.3 XP_006713601.1 2
XM_017005879.1 XP_016861368.1 2