gRNA Design Details
- Target DNA Sequence
- GGCAGTAGATGTCATTGTCT
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 79145670 - 79145692 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
83.5/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mfge8
- Entrez Gene ID
-
17304
- Gene Name
- milk fat globule-EGF factor 8 protein
- Gene Aliases
-
AA408458, AI325141, MFG-E8, MP47, Mfgm, P47, SED1, lactadherin
- Chromosome Location
-
Chr.7: 79133768 - 79149060 on GRCm38
- UniGene ID
- Mm.1451
- Species
-
Mouse