gRNA Design Details
- Target DNA Sequence
- GGATGGTCCTATTACAGCTT
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 116158286 - 116158308 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.5/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfbr2
- Entrez Gene ID
-
21813
- Gene Name
- transforming growth factor, beta receptor II
- Gene Aliases
-
1110020H15Rik, AU042018, DNIIR, RIIDN, TBR-II, TbetaR-II, TbetaRII
- Chromosome Location
-
Chr.9: 116087695 - 116175363 on GRCm38
- UniGene ID
- Mm.172346
- Species
-
Mouse