gRNA Design Details
- Target DNA Sequence
- GTGCTGCCAACAGGCGATAA
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 83667977 - 83667999 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
91.88/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ptger1
- Entrez Gene ID
-
19216
- Gene Name
- prostaglandin E receptor 1 (subtype EP1)
- Gene Aliases
-
42kDa, EP1, Ptgerep1
- Chromosome Location
-
Chr.8: 83666530 - 83670103 on GRCm38
- UniGene ID
- Mm.213000
- Species
-
Mouse