gRNA Design Details
- Target DNA Sequence
- GCCCAATAAAGGTTTTGCCA
- PAM Sequence
- AGG
- Target locus
-
Chr.3: 102514028 - 102514050 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.62/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ngf
- Entrez Gene ID
-
18049
- Gene Name
- nerve growth factor
- Gene Aliases
-
Ngfb
- Chromosome Location
-
Chr.3: 102469919 - 102521013 on GRCm38
- UniGene ID
- Mm.1259
- Species
-
Mouse