gRNA Design Details
- Target DNA Sequence
- CTGCCGGTACCATATGTAGG
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 48907354 - 48907376 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
88.5/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Htr3a
- Entrez Gene ID
-
15561
- Gene Name
- 5-hydroxytryptamine (serotonin) receptor 3A
- Gene Aliases
-
5-HT3
- Chromosome Location
-
Chr.9: 48899213 - 48911151 on GRCm38
- UniGene ID
- Mm.4831
- Species
-
Mouse