gRNA Design Details
- Target DNA Sequence
- CCTTCTTGTAGTTAGCCAGG
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 48908672 - 48908694 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Htr3a
- Entrez Gene ID
-
15561
- Gene Name
- 5-hydroxytryptamine (serotonin) receptor 3A
- Gene Aliases
-
5-HT3
- Chromosome Location
-
Chr.9: 48899213 - 48911151 on GRCm38
- UniGene ID
- Mm.4831
- Species
-
Mouse