gRNA Design Details
- Target DNA Sequence
- TCTGATGACTTCGTTGATGC
- PAM Sequence
- TGG
- Target locus
-
Chr.3: 87795060 - 87795082 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
82.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ntrk1
- Entrez Gene ID
-
18211
- Gene Name
- neurotrophic tyrosine kinase, receptor, type 1
- Gene Aliases
-
C80751, Tkr, TrkA, trk
- Chromosome Location
-
Chr.3: 87778244 - 87795242 on GRCm38
- UniGene ID
- Mm.80682
- Species
-
Mouse