gRNA Design Details
- Target DNA Sequence
- GATGGGAGCAACTAGAGATA
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 55760553 - 55760575 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
82.85/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgre4
- Entrez Gene ID
-
52614
- Gene Name
- adhesion G protein-coupled receptor E4
- Gene Aliases
-
D17Ertd479e, Egf-tm7, Emr4, Fire, Gpr127
- Chromosome Location
-
Chr.17: 55749984 - 55853662 on GRCm38
- UniGene ID
- Mm.210497
- Species
-
Mouse