gRNA Design Details
- Target DNA Sequence
- TGTGATCTTCCAGTCTGGGG
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 128729308 - 128729330 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.4/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mertk
- Entrez Gene ID
-
17289
- Gene Name
- c-mer proto-oncogene tyrosine kinase
- Gene Aliases
-
Eyk, Mer, Nyk, nmf12
- Chromosome Location
-
Chr.2: 128698997 - 128803162 on GRCm38
- UniGene ID
- Mm.239655
- Species
-
Mouse