gRNA Design Details
- Target DNA Sequence
- GCAAACGGTCTAGCTCACAG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 128248662 - 128248684 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.71/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tgfb1i1
- Entrez Gene ID
-
21804
- Gene Name
- transforming growth factor beta 1 induced transcript 1
- Gene Aliases
-
ARA55, Hic5, TSC-5, hic-5
- Chromosome Location
-
Chr.7: 128246799 - 128255699 on GRCm38
- UniGene ID
- Mm.3248
- Species
-
Mouse