gRNA Design Details
- Target DNA Sequence
- CATCCTGGAGATTACTGCGG
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 144840672 - 144840694 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
96.98/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Fgf3
- Entrez Gene ID
-
14174
- Gene Name
- fibroblast growth factor 3
- Gene Aliases
-
Fgf-3, Int-2, Int-P
- Chromosome Location
-
Chr.7: 144837521 - 144844447 on GRCm38
- UniGene ID
- Mm.4947
- Species
-
Mouse