gRNA Design Details

Target DNA Sequence
CAGGTCTAAACGTTCGGGGC
PAM Sequence
TGG
Target locus
Chr.9: 64020997 - 64021019 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 92.36/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Smad6
Entrez Gene ID
17130
Gene Name
SMAD family member 6
Gene Aliases
Madh6, b2b390Clo
Chromosome Location
Chr.9: 63953076 - 64022085 on GRCm38
UniGene ID
Mm.325757
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_008542.3 NP_032568.3 1