gRNA Design Details

Target DNA Sequence
CATAGTAGTACCGCAAGGCC
PAM Sequence
CGG
Application
Gene Knock-Out
Score: 75.16/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
ELK1
Entrez Gene ID
2002
Gene Name
ELK1, ETS transcription factor
Gene Aliases
-
Chromosome Location
Chr.X: 47635520 - 47650604 on GRCh38
UniGene ID
Hs.181128
Species
Human

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001114123.2 NP_001107595.1 3
NM_001257168.1 NP_001244097.1 3
NM_005229.4 NP_005220.2 2
XM_017029339.1 XP_016884828.1 1