gRNA Design Details
- Target DNA Sequence
- GTGTCCTACCCAGAGGTTGC
- PAM Sequence
- AGG
- Application
- Gene Knock-Out
Score:
84.84/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Elk1
- Entrez Gene ID
-
13712
- Gene Name
- ELK1, member of ETS oncogene family
- Gene Aliases
-
Elk-1
- Chromosome Location
-
Chr.X: 20933395 - 20950608 on GRCm38
- UniGene ID
- Mm.405823
- Species
-
Mouse