gRNA Design Details
- Target DNA Sequence
- AGCACTTCTGCTCATCCGCG
- PAM Sequence
- TGG
- Target locus
-
Chr.6: 87845180 - 87845202 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.44/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Cnbp
- Entrez Gene ID
-
12785
- Gene Name
- cellular nucleic acid binding protein
- Gene Aliases
-
AA408710, Cnbp1, Znf9
- Chromosome Location
-
Chr.6: 87842615 - 87851106 on GRCm38
- UniGene ID
- Mm.290251
- Species
-
Mouse