gRNA Design Details
- Target DNA Sequence
- ACTTCATCAATTGGAGGATT
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 36012895 - 36012917 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
83.86/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Kpna1
- Entrez Gene ID
-
16646
- Gene Name
- karyopherin (importin) alpha 1
- Gene Aliases
-
AW494490, IPOA5, NPI1, Rch2, mSRP1
- Chromosome Location
-
Chr.16: 35983363 - 36037136 on GRCm38
- UniGene ID
- Mm.6952
- Species
-
Mouse