gRNA Design Details
- Target DNA Sequence
- CGAGGATGTGCCTGAAGGTC
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 35588585 - 35588607 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mfhas1
- Entrez Gene ID
-
52065
- Gene Name
- malignant fibrous histiocytoma amplified sequence 1
- Gene Aliases
-
2310066G09Rik, D8Ertd91e
- Chromosome Location
-
Chr.8: 35587777 - 35679449 on GRCm38
- UniGene ID
- Mm.103246
- Species
-
Mouse