gRNA Design Details

Target DNA Sequence
TATGAATGTGTCTCCTCGAT
PAM Sequence
AGG
Target locus
Chr.13: 89042486 - 89042508 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 88.18/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Edil3
Entrez Gene ID
13612
Gene Name
EGF-like repeats and discoidin I-like domains 3
Gene Aliases
Del-1, Del1
Chromosome Location
Chr.13: 88821472 - 89323225 on GRCm38
UniGene ID
Mm.125580
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001037987.3 NP_001033076.1 4
NM_010103.4 NP_034233.1 3