gRNA Design Details
- Target DNA Sequence
- GTTGGCCGACTGAGACTTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 6624123 - 6624145 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.48/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tbrg4
- Entrez Gene ID
-
21379
- Gene Name
- transforming growth factor beta regulated gene 4
- Gene Aliases
-
2310042P22Rik, AA120735, AA408001, AI527316, Cpr2, R74877, Tb-12, mKIAA0948
- Chromosome Location
-
Chr.11: 6615598 - 6626084 on GRCm38
- UniGene ID
- Mm.247761
- Species
-
Mouse