gRNA Design Details
- Target DNA Sequence
- AGTGGTCAACTCGGAGTCTC
- PAM Sequence
- CGG
- Target locus
-
Chr.3: 95738060 - 95738082 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.92/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ecm1
- Entrez Gene ID
-
13601
- Gene Name
- extracellular matrix protein 1
- Gene Aliases
-
AI663821, p85
- Chromosome Location
-
Chr.3: 95734147 - 95739819 on GRCm38
- UniGene ID
- Mm.3433
- Species
-
Mouse