gRNA Design Details

Target DNA Sequence
TCGTACACAGGGATGTCTTC
PAM Sequence
TGG
Target locus
Chr.3: 95737849 - 95737871 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 85.31/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Ecm1
Entrez Gene ID
13601
Gene Name
extracellular matrix protein 1
Gene Aliases
AI663821, p85
Chromosome Location
Chr.3: 95734147 - 95739819 on GRCm38
UniGene ID
Mm.3433
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001252653.1 NP_001239582.1 4
NM_007899.2 NP_031925.2 4